missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk pack
Unit Size:
Pack of 108
Pack of 12
Pack of 160
Pack of 24
Pack of 288
Pack of 48
Description
Ideal for solvent, chemical and sample storage
- Narrow mouth enables liquids to be carefully poured to prevent spillage
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
- Available in partitioned Convenience or Valu-Bulk™ cartons
Order Info
Caps can be ordered separately.
Compliance
USP Type-III soda-lime glass
Specifications
Specifications
| Capacity | 8 oz., 250 mL |
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8 oz. |
| Color | Clear |
| Diameter (English) | 2.36 in. |
| Material | Soda Lime Glass |
| Packaging | Valu-bulk pack |
| Closure Color | White |
| Certifications/Compliance | EPA, FDA |
| Height (Metric) | 138 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction