missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps

Product Code. 10133342 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience Pack
Valu-bulk Pack
Unit Size:
Pack of 12
Pack of 144
Pack of 24
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging Unit Size
10133342 1000 mL Convenience Pack Pack of 12
10425595 60 mL Valu-bulk Pack Pack of 144
10717944 60 mL Convenience Pack Pack of 24
10386942 125 mL Convenience Pack Pack of 24
10051332 250 mL Convenience Pack Pack of 12
10768324 500 mL Convenience Pack Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10133342 Supplier DWK Life Sciences Supplier No. W216907
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for soil sampling and environmental applications

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas

Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage

Order Info

Caps can be ordered separately.

Compliance

Clear-USP Type III soda-lime glass

Specifications

Capacity (Metric) 1000 mL
Color Clear
Diameter (English) 3.74 in.
Material Glass
Packaging Convenience Pack
Closure Color White
Height (Metric) 170 mm
Height (English) 6.69 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 89-400 mm
Diameter (Metric) 95 mm
Closure Type Screw Cap
Capacity (English) 32 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.