Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Shop All Wheaton Science Products ProductsDescription
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
Clear-USP Type III soda-lime glass
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Clear |
| Diameter (English) | 3.58 in. |
| Material | Glass |
| Packaging | Convenience Pack |
| Closure Color | White |
| Height (Metric) | 95 mm |
| Height (English) | 3.7 in. |
| Closure Liner | Poly/Vinyl |
| Closure Material | Polypropylene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.