missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK™ Amber Kits

Product Code. 17992641 Shop All Wheaton Science Products Products
Change view
Click to view available options
Closure Color:
Blue
Red
Closure Type:
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure Serum Stopper
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
1 set
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Closure Type Volume (Metric) Unit Size
17992641 Red OmniFlex 3G Serum Stopper 5 mL 1 set
17912641 Blue OmniFlex 3G Igloo Lyo Stopper 2 mL 1 set
17952641 Red OmniFlex 3G Igloo Lyo Stopper 2 mL 1 set
17932581 Blue OmniFlex 3G Serum Stopper 2 mL 1 set
17962641 Red OmniFlex 3G Serum Stopper 2 mL 1 set
17922641 Blue Ultra Pure Serum Stopper 2 mL 1 set
17932641 Red Ultra Pure Serum Stopper 2 mL 1 set
17932591 Blue OmniFlex 3G Igloo Lyo Stopper 5 mL 1 set
17912651 Red OmniFlex 3G Igloo Lyo Stopper 5 mL 1 set
17982641 Blue OmniFlex 3G Serum Stopper 5 mL 1 set
17992581 Blue Ultra Pure Serum Stopper 5 mL 1 set
17972641 Red Ultra Pure Serum Stopper 5 mL 1 set
17902651 Blue OmniFlex 3G Igloo Lyo Stopper 10 mL 1 set
17942651 Red OmniFlex 3G Igloo Lyo Stopper 10 mL 1 set
17962591 Blue OmniFlex 3G Serum Stopper 10 mL 1 set
17932651 Red OmniFlex 3G Serum Stopper 10 mL 1 set
17922651 Blue Ultra Pure Serum Stopper 10 mL 1 set
17962571 Red Ultra Pure Serum Stopper 10 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 17992641 Supplier DWK Life Sciences Supplier No. W991010CPA
  • ONLINE EXCLUSIVE PRICE
    1575.00 €
    1510.27 € / 1 sets
    Save 64.73 €
    4% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions

Specifications

Closure Color Red
Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Volume (Metric) 5 mL
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Graduated No
Includes Vial, Stopper, and Seal
No. per Case 120/Cs.
Style Crimp top
Product Line Wheaton CompletePAK
Closure Type OmniFlex 3G Serum Stopper
Closure Size 20 mm
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 40 mm
Product Type COMPLETEPAK Amber
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.