missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits

Product Code. 16297460 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
Omni Flex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Sterile
OmniFlex 3G Sterile Igloo Lyo Stopper
OmniFlex 3G Sterile Serum Stopper
Ultra Pure Sterile
Ultra Pure Sterile Serum Stopper
Color (Seal):
Blue
Red
Unit Size:
Pack of 158
Pack of 200
Pack of 210
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Closure Type Color (Seal) Unit Size
16297460 2 mL Ultra Pure Sterile Blue Pack of 210
16227300 2 mL Omni Flex 3G Blue Pack of 210
16237300 2 mL OmniFlex 3G Sterile Blue Pack of 210
16247300 2 mL Ultra Pure Sterile Red Pack of 210
16257300 2 mL Omni Flex 3G Red Pack of 210
16267300 2 mL OmniFlex 3G Sterile Red Pack of 210
16277300 5 mL Ultra Pure Sterile Serum Stopper Blue Pack of 200
16287300 5 mL OmniFlex 3G Sterile Serum Stopper Blue Pack of 200
16297300 5 mL OmniFlex 3G Igloo Lyo Stopper Blue Pack of 200
16207310 5 mL Ultra Pure Sterile Serum Stopper Red Pack of 200
16217310 5 mL OmniFlex 3G Sterile Serum Stopper Red Pack of 200
16227310 5 mL OmniFlex 3G Sterile Igloo Lyo Stopper Red Pack of 200
16237310 10 mL Ultra Pure Sterile Serum Stopper Blue Pack of 158
16247310 10 mL OmniFlex 3G Sterile Serum Stopper Blue Pack of 158
16257310 10 mL OmniFlex 3G Sterile Igloo Lyo Stopper Blue Pack of 158
16267310 10 mL Ultra Pure Sterile Serum Stopper Red Pack of 158
16277310 10 mL OmniFlex 3G Sterile Serum Stopper Red Pack of 158
16287310 10 mL OmniFlex 3G Sterile Igloo Lyo Stopper Red Pack of 158
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 16297460 Supplier DWK Life Sciences Supplier No. W990000CP

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications

Color (Seal) Blue
Color Clear
Material Glass
Sterility Sterile
Volume (Metric) 2 mL
Includes Vial, Stopper, and Seal
No. per Case 210
Product Line Wheaton CompletePAK
Volume (Metric) Total 2 mL
Closure Material Rubber (Stopper)
Closure Type Ultra Pure Sterile
Quantity 210 Cs.
Closure Size 13 mm
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Type Sterile Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.