Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
Product Code. 17912611 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Amber
Clear
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 104
Pack of 120
Pack of 278
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) unitSize
17912611 Clear 10 mL Pack of 104
17922611 Amber 2 mL Pack of 278
17942611 Amber 5 mL Pack of 120
17912581 Amber 10 mL Pack of 104
17922581 Clear 2 mL Pack of 278
17902681 Clear 5 mL Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17912611 Supplier DWK Life Sciences Supplier No. W987002S
Only null left
Add to basket
Add to basket
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
Certifications
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Borosilicate Glass (Type 1)
Sterility Sterile
Volume (Metric) 10 mL
No. per Case 104/Cs.
Product Line Wheaton CompletePAK
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified