Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
Product Code. 16297280 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Clear
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 158
Pack of 200
Pack of 210
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Unit Size
16297280 Clear 2 mL Pack of 210
16207290 Clear 5 mL Pack of 200
16217290 Clear 10 mL Pack of 158
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16297280 Supplier DWK Life Sciences Supplier No. W986000S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Color Clear
Material Borosilicate Glass (Type 1)
Sterility Sterile
Volume (Metric) 2 mL
No. per Case 210
Product Line Wheaton CompletePAK
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified