missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Wheaton™ Cover for Glass Staining Dish designed for use with Wheaton glass staining dishes
- Manufactured from soda-lime glass for durability and chemical resistance
- Helps protect slides and staining solutions during manual staining procedures
- Available in configurations compatible with multiple slide rack sizes
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Bottom Shape | Flat |
| Color | Clear |
| Material | Soda Lime Glass |
| Type | Mix and Match Cover |
| For Use With (Equipment) | Glass Staining Dish for 16, 30 and 40 Slide Racks |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?