Learn More
DWK Life Sciences WHEATON™ Glass Slide Staining Rack for 16 Slides
Product Code. 10061392 Shop All Wheaton Science Products Products
Change view
Click to view available options
Slide Count:
16 Slides or 32 Slides Back to Back
Unit Size:
Pack of 3
Product Code. Slide Count Unit Size
10061392 16 Slides or 32 Slides Back to Back Pack of 3
1 options
Product Code. 10061392 Supplier DWK Life Sciences Supplier No. 900254
  • ONLINE EXCLUSIVE PRICE
    236.00 €
    215.75 € / Pack of 3
    Save 20.25 €
    8% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WHEATON™ Glass Slide Staining Rack for 16 Slides is manufactured from soda-lime glass and holds up to 16 slides. Designed for manual staining procedures, it features an open bottom to facilitate rapid immersion and drainage, reducing carryover.
Made of Soda-lime glass
  • Wheaton™ Glass Slide Staining Rack designed for manual staining procedures
  • Manufactured from soda-lime glass for chemical resistance and durability
  • Holds up to 16 slides
  • Open bottom design facilitates rapid immersion and drainage, reducing carryover
  • Compatible with standard slide sizes: 3 x 1 in. (75 x 25 mm), 3 x 1.5 in. (75 x 38 mm), and 3 x 2 in. (75 x 51 mm)
Specifications
Autoclavable Not Autoclavable
Color Clear
Material Soda Lime Glass
Type Slide Rack
Handle Type Hinged
Slide Count 16 Slides or 32 Slides Back to Back