missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ HDPE Serum Bottles

Product Code. 12791588 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
Unit Size:
Pack of 100
Pack of 200
Pack of 300
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12791588 120 mL Pack of 100
12807632 15 mL Pack of 300
12422621 30 mL Pack of 200
12432621 60 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12791588 Supplier DWK Life Sciences Supplier No. 224037
  • ONLINE EXCLUSIVE PRICE
    253.00 €
    228.24 € / Pack of 100
    Save 24.76 €
    9% Off
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wheaton Serum bottle is manufactured from HDPE and widely used for animal health products and storage of non-medical products.

  • Manufactured from high density polyethylene
  • Widely used for animal health products and storage of non-medical products
  • Accepts standard rubber stoppers and aluminum seals

Specifications

Capacity 4.05 oz., 120 mL
Capacity (Metric) 120 mL
Color Natural
Diameter (English) 1.85 in.
Diameter (English) Inner Neck 0.78 in.
Diameter (Metric) Outer Neck 20 mm
Diameter (Metric) Inner Neck 13 mm
Material HDPE
Height (Metric) 103 mm
Height (English) 4.05 in.
Closure Material Rubber
Diameter (Metric) 47 mm
Capacity (English) 4.05 oz.
Product Type Serum Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.