missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Labeled Reagent Bottles

Product Code. 12452541 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12452541 250 mL Pack of 6
12462541 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12452541 Supplier DWK Life Sciences Supplier No. 220223
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Autoclavable Reagent bottle manufactured from USP Type I borosilicate glass and features no-drip pour lip

  • Pre-attached polypropylene screw cap with PTFE liner
  • Large writing patch
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.45 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Diameter (English) 2.63 in.
Includes No-drip pour lip, large writing patch
Material Glass
Mouth Wide
Height (Metric) 150 mm
Product Type Reagent Bottle
Height (English) 5.9 in.
Closure Liner Polyethylene
Closure Material Polypropylene
Diameter (Metric) 67 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.