missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Labeled Reagent Bottles

Product Code. 12462541 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12462541 500 mL Pack of 6
12452541 250 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12462541 Supplier DWK Life Sciences Supplier No. 220283
  • ONLINE EXCLUSIVE PRICE
    139.00 €
    117.52 € / Pack of 6
    Save 21.48 €
    15% Off
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ Labeled Reagent Bottles are autoclavable and manufactured from USP Type I borosilicate glass. Featuring a no-drip pour lip, pre-attached polypropylene screw cap with PTFE liner, and a large writing patch for easy labeling.

  • Manufactured from USP Type I borosilicate glass for superior chemical resistance
  • Autoclavable for convenient sterilization
  • No-drip pour lip for clean, aseptic pouring
  • Pre-attached polypropylene screw cap with PTFE liner for a secure, leak-resistant seal
  • Large writing patch for easy sample identification and labeling

Specifications

Capacity 16.9 oz., 500mL
Capacity (Metric) 500 mL
Color Clear
Diameter (English) 3.46 in.
Includes No-drip pour lip, large writing patch
Material Borosilicate Glass
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 190 mm
Height (English) 7.48 in.
Chemical Label Unlabeled
Closure Liner Polyethylene
Closure Material Polypropylene
Diameter (Metric) 88 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Capacity (English) 16.9 oz.
Shape Round
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.