missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Labeled Reagent Bottles

Product Code. p-45330 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12452541

Brand: DWK Life Sciences 220223

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Autoclavable Reagent bottle manufactured from USP Type I borosilicate glass and features no-drip pour lip

  • Pre-attached polypropylene screw cap with PTFE liner
  • Large writing patch
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.45 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Diameter (English) 2.63 in.
Includes No-drip pour lip, large writing patch
Material Glass
Mouth Wide
Height (Metric) 150 mm
Product Type Reagent Bottle
Height (English) 5.9 in.
Closure Liner Polyethylene
Closure Material Polypropylene
Diameter (Metric) 67 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.