Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (13mm)
13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
Brand: DWK Life Sciences 242412
Code : A0
Additional Details : Weight : 0.47630kg
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 100
Specifications
Red | |
100 Cs. | |
ISO 9001 | |
Plastic dropping bottle |
LDPE | |
Dropper Tips | |
41 μL |