missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories

Product Code. 11718719 Shop All Wheaton Science Products Products
Change view
Click to view available options
Product Type:
Index Card
M-T Vial File
Partition
Storage Box
Quantity:
35 Case
6 Case
For Use With (Equipment):
228778 Hinged Case
228780 Hinged Case
24 x 20mL vials
40 x 4mL vials
54 x 8mL vials
60 x 2mL vials
W228790 Hinged Case
W228792 Hinged Case
Unit Size:
Pack of 35
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity For Use With (Equipment) unitSize
11718719 M-T Vial File 6 Case 60 x 2mL vials Pack of 6
10556525 Index Card 35 Case 228780 Hinged Case Pack of 35
10116834 Partition 6 Case 228778 Hinged Case Pack of 6
12453761 Index Card 35 Case 228778 Hinged Case Pack of 35
10362663 Storage Box 6 Case 40 x 4mL vials Pack of 6
15880130 Storage Box 6 Case 54 x 8mL vials Pack of 6
13429698 Storage Box 6 Case 24 x 20mL vials Pack of 6
15343198 Partition 6 Case W228792 Hinged Case Pack of 6
15843731 Partition 6 Case W228790 Hinged Case Pack of 6
10164613 Partition 6 Case 228780 Hinged Case Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11718719 Supplier DWK Life Sciences Supplier No. 228778
  • ONLINE EXCLUSIVE PRICE
    192.50 €
    189.96 € / Pack of 6
    Save 2.54 €
    1% Off
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For compact and organized sample storage

  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polystyrene
For Use With (Application) Vial storage and transport
For Use With (Equipment) 60 x 2mL vials
Includes Clear polystyrene hinged case, partitions, foam inserts and alphanumerical indexing card
No. of Places 60
Product Type M-T Vial File
Quantity 6 Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.