missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Polypropylene Serum Bottles

Product Code. 12694783 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
3 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12694783 3 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12694783 Supplier DWK Life Sciences Supplier No. 224007
  • ONLINE EXCLUSIVE PRICE
    367.00 €
    344.68 € / Pack of 500
    Save 22.32 €
    6% Off
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ Polypropylene Serum Bottles are manufactured from chemical resistant polypropylene, accept standard rubber stoppers and aluminum seals, and are autoclavable at 121°C for 20 minutes. Available in a 3mL size, ideal for sample storage and diluent solutions.

  • Manufactured from chemical resistant polypropylene
  • Accepts standard rubber stoppers and aluminum seals
  • Autoclavable at 121°C for 20 minutes
  • Available in 3mL size
  • Suitable for diluent solutions and sample storage applications
  • Available in bulk quantity configurations — contact your DWK Sales Representative

Specifications

Capacity (Metric) 3 mL
Color Natural
Diameter (English) 0.66 in.
Diameter (English) Inner Neck 0.51 in.
Diameter (Metric) Outer Neck 13mm
Diameter (Metric) Inner Neck 7mm
Material Polypropylene
Height (Metric) 38 mm
Height (English) 1.49 in.
Closure Material Rubber
Diameter (Metric) 17 mm
Product Type Serum Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.