missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers

Product Code. 11734219 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
15-425
20-400
22-400
24-400
28-400
33-400
Unit Size:
Pack of 144
This item is not returnable. View return policy

Product Code. 11734219

Brand: DWK Life Sciences 239257

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Unique cone liner design offers superior torque retention and excellent sealing characteristics

  • Poly-Seal liner is manufactured from low density polyethylene
  • Phenolic
  • Screw cap
  • Color: Black

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 24-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ container
Closure Liner Polyseal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.