missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Serum Bottles and Vials

Product Code. 10768134 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
60 mL
100 mL
125 mL
Color:
Amber
Clear
No. per Pack:
144/Pk.
288/Pk.
Diameter (Metric):
15.6 x 36 mm
23 x 47 mm
25 x 54 mm
32 x 58 mm
37 x 62 mm
37 x 63 mm
41 x 91 mm
43 x 73 mm
52 x 95 mm
54 x 107 mm
Unit Size:
Pack of 144
Pack of 288
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color No. per Pack Diameter (Metric) Unit Size
10768134 10 mL Clear 288/Pk. 25 x 54 mm Pack of 288
11591165 2 mL Clear 288/Pk. 15.6 x 36 mm Pack of 288
10520853 5 mL Clear 288/Pk. 23 x 47 mm Pack of 288
10424292 5 mL Amber 288/Pk. 23 x 47 mm Pack of 288
10434292 10 mL Amber 288/Pk. 25 x 54 mm Pack of 288
10453712 20 mL Clear 288/Pk. 32 x 58 mm Pack of 288
10600173 20 mL Amber 288/Pk. 32 x 58 mm Pack of 288
10071392 30 mL Amber 288/Pk. 37 x 62 mm Pack of 288
10632973 30 mL Clear 288/Pk. 37 x 63 mm Pack of 288
10464672 50 mL Amber 288/Pk. 43 x 73 mm Pack of 288
10642973 50 mL Clear 288/Pk. 43 x 73 mm Pack of 288
10235262 60 mL Clear 144/Pk. 41 x 91 mm Pack of 144
10011422 100 mL Amber 144/Pk. 52 x 95 mm Pack of 144
10021362 100 mL Clear 144/Pk. 52 x 95 mm Pack of 144
10071382 125 mL Clear 144/Pk. 54 x 107 mm Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 10768134 Supplier DWK Life Sciences Supplier No. 223739
  • ONLINE EXCLUSIVE PRICE
    460.00 €
    393.48 € / Pack of 288
    Save 66.52 €
    14% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bottles offer exceptional wall strength and superior heat and chemical tolerances

  • Ideal for long- and short-term sample storage and vaccine/injectable drug containers
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage

Compliance

Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

Specifications

Capacity 0.34 oz., 10 mL
Capacity (Metric) 10 mL
Color Clear
Diameter (English) 0.98 in.
Material Borosilicate Glass
No. per Pack 288/Pk.
Quantity 288/Pk.
Sterility Non-sterile
Mouth Narrow
Height (Metric) 54 mm
Height (English) 2.12 in.
Diameter (Metric) 25 x 54 mm
Capacity (English) 0.34 oz.
Shape Round
Product Type Serum Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.