missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
209.00 € - 376.00 €
Product Code | Brand | Color | Product Type | For Use With (Equipment) | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | For Use With (Equipment) | For Use With (Application) | Price | Quantity & Availability | ||
11764189
|
DWK Life Sciences
W224100-093 |
Gray | 2-Leg Lyophilization Stopper; Butyl Rubber | Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials | Lyophilization |
209.00 €
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||
10021492
|
DWK Life Sciences
224100-172 |
Red | Straight-plug style; Rubber | Fits 1.3mm I.D., 20mm O.D. mouth of serum bottles and vials | Not for lyophilization |
273.00 €
Pack of 1000 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
10452242
|
DWK Life Sciences
224100-192 |
Gray | 2-Leg Lyophilization Stopper; Chlorobutyl Isoprene blend | Fits 13mm I.D., 20mm O.D. mouth of serum bottles and vials | Lyophilization |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Provide airtight covers
- Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
- Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
Yes |