missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Straight Plug Rubber Stoppers

Product Code. 12432631 Shop All Wheaton Science Products Products
Click to view available options
Color:
Gray
Red
Diameter (Metric) Outer:
13 mm
20 mm
Material:
Chlorobutyl-Isoprene
FEP
Rubber
Product Type:
Straight Plug Stopper
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12432631

Brand: DWK Life Sciences 224100170

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides maximum sealing for vials with straight-wall glass finishes. Wheaton™ Straight Plug Rubber Stoppers are manufactured from the highest quality raw materials.

  • Autoclavable
  • Available in a variety of sizes/colors
  • Manufactured to yield exceptional dimensional stability
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Vacule Ampules
Color Red
Diameter (Metric) Outer 20 mm
Material Rubber
Product Type Straight Plug Stopper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.