missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners

Product Code. 10210332 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
15-415
20-410
24-400
24-410
28-400
28-410
33-400
33-430
Liner Material:
Foam/LDPE
Polyethylene Foam
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 10210332

Brand: DWK Life Sciences 242220

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For general storage and packaging applications

  • Lined with foam polyethylene
  • Excellent barrier to aqueous solutions
  • Repeat sealability

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 200 Cs.
Liner Material Polyethylene Foam
Neck Finish 24-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.