missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
15-415
20-410
24-400
24-410
28-400
28-410
33-400
33-430
Liner Material:
Foam/LDPE
Polyethylene Foam
Unit Size:
Pack of 200
Description
- Lined with foam polyethylene
- Excellent barrier to aqueous solutions
- Repeat sealability
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 200 Cs. |
| Liner Material | Polyethylene Foam |
| Neck Finish | 15-415 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Glass bottle or vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction