missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners

Product Code. 11748829 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
22-400
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
89-400
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
This item is not returnable. View return policy

Product Code. 11748829

Brand: DWK Life Sciences 239213

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products

  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 38-400
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Round bottle
Closure Liner Polyvinyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.