missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners

Product Code. 11794219 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Liner Material:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
Polypropylene, PTFE
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Liner Material Material No. per Case Unit Size
11794219 22-400 Polyethylene Foam Polypropylene 144 Cs. Pack of 144
11784219 20-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11714229 28-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11708819 63-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11798809 53-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11704229 24-400 PTFE/Polyethylene Polypropylene, PTFE 144 Cs. Pack of 144
11778809 43-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11788809 45-400 Polyethylene Foam Polypropylene 72 Cs. Pack of 72
11724229 33-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11734229 48-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11744229 58-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11768809 38-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
10113052 89-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11754229 70-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 11794219 Supplier DWK Life Sciences Supplier No. 239231
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    85.75 € / Pack of 144
    Save 35.25 €
    29% Off
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners seal glass bottles and vials securely. The PTFE-faced foamed PE liner resists chemicals and compresses for a reliable seal, ideal for environmental samples, pharmaceuticals, and diagnostic reagents.

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE-faced foamed polyethylene liner offers chemical resistance against high purity chemicals, strong acids, and solvents
  • Polyethylene foam provides compressibility and reliable sealing properties
  • Suitable for environmental samples, pharmaceuticals, and diagnostic reagents
  • Compatible with glass bottles and vials
  • White polypropylene screw cap construction
  • Available in multiple GPI cap finish sizes to fit a range of bottle and vial neck finishes

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Sterility Non-sterile
Liner Material Polyethylene Foam
Neck Finish 22-400
For Use With (Equipment) Glass bottle or vial
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.