missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ White Acetal Copolymer Hose Clamp

Product Code. 11759899
Change view
Click to view available options
For Use With:
0.27 in. x 0.29 in. Tubing I.D.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11759899 0.27 in. x 0.29 in. Tubing I.D. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11759899 Supplier Masterflex™ Supplier No. 9561400
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Material Acetal Copolymer
For Use With 0.27 in. x 0.29 in. Tubing I.D.
Packaging Quantity 100 Pack
Color White
Product Type Hose Clamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.