Learn More
3M™ White/Blue Combined SMS Laminate Protective Coveralls
Product Code. 12962325 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12962325 3X-Large Pack of 20
12912325 Small Pack of 20
12922325 Medium Pack of 20
12932325 Large Pack of 20
12942325 X-Large Pack of 20
11963784 2X-Large Pack of 20
12952325 2X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12962325 Supplier 3M™ Supplier No. 7000089640
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Protective coveralls are manufactured from a high quality laminated material with a breathable back panel to improve air ventilation and reduce heat build-up.
  • Breathable back panel to help improve air ventilation and reduce heat build-up.
  • Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
  • Elastic waist and ankles for convenience and freedom of movement.
  • Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Neck Style Hooded
Sterility Non-sterile
Protection Class Category III
Model 4535W
Height (Metric) 194 to 200 cm