missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ White Coverall
BIOPHARMA

Product Code. 15258128
Change view
Click to view available options
Size:
2X-Large
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15258128 2X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15258128 Supplier Nitritex™ Supplier No. BDCHT2XL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles and thumb loops to ensure a secure hold. The BioClean-D coverall provides exceptional comfort and protection, even when handling cytotoxic drugs

Cuffs & ankles ISO Class 4 cleanroom compatible zip with adhesive cover flap, Type 5 & 6 White 2XL BioClean-D
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Color White
Disposable Single-use
Sterility Non-sterile
Size (Metric) Chest 116 to 124 cm
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.