missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ White Coverall, XS
BIOPHARMA

Product Code. 12727196
Change view
Click to view available options
Size:
X-Small
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12727196 X-Small Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12727196 Supplier Nitritex™ Supplier No. BDCHTXS
Add to basket
Add to basket
This item is not returnable. View return policy

Cleanroom compatible, non-sterile coverall

Cuffs & ankles ISO Class 4 cleanroom compatible zip with adhesive cover flap, Type 5 & 6 White XS BioClean-D
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size X-Small
Color White
Disposable Single-use
Sterility Non-sterile
Size (Metric) Chest 76 to 84 cm
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.