Learn More
Buerkle™ White Disposable Forceps
BIOPHARMAGREENER_CHOICE
Product Code. 16505210 Shop All Bürkle Products
Change view
Click to view available options
Tip Style:
Angled, Pointed
Broad
Pointed
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style Sterility unitSize
16505210 Angled, Pointed Non-sterile Pack of 100
16515210 Pointed Non-sterile Pack of 100
16525210 Broad Non-sterile Pack of 100
16535210 Angled, Pointed Sterile Pack of 100
16545210 Pointed Sterile Pack of 100
16555210 Broad Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16505210 Supplier Buerkle™ Supplier No. 53780040
  • ONLINE EXCLUSIVE PRICE
    49.15 €
    37.25 € / Pack of 100
    Save 11.90 €
    24% Off
In Stock
Add to basket
Add to basket
Disposable forceps are ideal for grasping, lifting, and moving small objects like fine tissue or solid samples.
  • The fluted handles ensure optimum and secure handling as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Sterile forceps are individually wrapped
  • For single use
  • Cleanroom manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
TRUSTED_SUSTAINABILITY
Specifications
Product Type Forceps
Length (English) 5.12 in.
Length (Metric) 130 mm
Material Polystyrene
No. per Pack 100 (large pack)
Sterility Non-sterile
Style Standard
Tip Style Angled, Pointed