missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Visitor Lab Coat, Series 4400

Product Code. 12857310
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12857310 4X-Large Pack of 50
12951335 Medium Pack of 50
12931335 Small Pack of 50
12847310 3X-Large Pack of 50
12921335 2X-Large Pack of 50
12981325 Large Pack of 50
12901335 X-Large Pack of 50
12941335 Small Pack of 50
15351495 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12857310 Supplier 3M™ Supplier No. 7000089699
  • ONLINE EXCLUSIVE PRICE
    280.00 €
    168.69 € / Pack of 50
    Save 111.31 €
    39% Off
View all promotions
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Offers a barrier against non-hazardous particles and liquid splashes. The Series 4400 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.

  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Elastic cuffs
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Resistance Non-hazardous Liquids, Dust
Seam Type Serged
Size (English) Chest 46 to 52 in.
Size (Metric) Chest 117 to 132 cm
Certifications/Compliance (Europe) CE Category 1
For Use With (Application) Non-hazardous liquids and dust
Includes Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
Packaging Type Individually Packaged
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.