missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Plastic General Purpose Trays

Product Code. 12347237 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
460 x 260mm
570 x 340mm
570 x 540mm
680 x 540mm
700 x 460mm
1130 x 540mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12347237 460 x 260mm Each
12357237 570 x 340mm Each
12367237 570 x 540mm Each
12377237 680 x 540mm Each
12387237 700 x 460mm Each
12397237 1130 x 540mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12347237
Estimated Shipment: 19-08-2026
Add to basket
Add to basket

Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25

Specifications

Dimensions (L x W) 460 x 260mm
Color White
Length (Metric) 460 mm
Material Plastic
Type Tray
Width (Metric) 260 mm
For Use With Safe containment of spillages, lab bench protection
Includes Liner
No. per Pack 1 Each
Shape Rectangular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.