Learn More
Fisherbrand™ White Polypropylene Caps
Product Code. 16134607 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Liner Material:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
Unit Size:
Pack of 100
Pack of 1300
Pack of 144
Pack of 2100
Pack of 288
Pack of 370
Pack of 500
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Liner Material Quantity unitSize
16134607 53-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16408125 33-400 PTFE/Polyethylene Foam 2100/Cs. Pack of 2100
16164477 53-400 Pulp/Vinyl 1300/Cs. Pack of 1300
16134597 18-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16124597 15-425 PTFE/Polyethylene Foam 500/Cs. Pack of 500
16144597 20-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16164597 24-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16154597 22-400 PTFE/Polyethylene Foam 500/Cs. Pack of 500
16174597 28-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16184597 33-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16194597 38-400 PTFE/Polyethylene Foam 500/Cs. Pack of 500
16104607 43-400 PTFE/Polyethylene Foam 500/Cs. Pack of 500
16114607 45-400 PTFE/Polyethylene Foam 500/Cs. Pack of 500
16124607 48-400 PTFE/Polyethylene Foam 100/Cs. Pack of 100
16144607 58-400 PTFE/Polyethylene Foam 100/Cs. Pack of 500
16154607 70-400 PTFE/Polyethylene Foam 144/Cs. Pack of 144
16164607 83-400 PTFE/Polyethylene Foam 144/Cs. Pack of 144
16174607 89-400 PTFE/Polyethylene Foam 144/Cs. Pack of 144
16184607 63-400 PTFE/Polyethylene Foam 288/Cs. Pack of 288
16478125 53-400 Polyethylene Foam 1300/Cs. Pack of 1300
16124617 53-400 Polyethylene Foam 500/Cs. Pack of 500
16488125 89-400 Polyethylene Foam 370/Cs. Pack of 370
16114647 53-400 Pulp/Vinyl 288/Cs. Pack of 288
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 16134607
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
White polypropylene screw caps provides clear surface for writing on cap
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color White
Material Polypropylene
Quantity 100/Cs.
Liner Material PTFE/Polyethylene Foam
Neck Finish 53-400
Product Type Lined Cap