Learn More
Fisherbrand™ White Polypropylene Caps
Shop All Fisherbrand Products
Click to view available options
Neck Finish:
13-425
48-400
70-400
8-425
Product Type:
Lined Cap
Unlined Cap
Unit Size:
Pack of 144
Pack of 1700
Pack of 500
Pack of 576
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 576 |
| Liner Material | Pulp/Vinyl |
| Neck Finish | 48-400 |
| Product Type | Lined Cap |