missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Sterile cover for smart phones
- Constructed of ultra-clear polyethylene for clear photos and is touchscreen compatible with gloves
- Protects the sterile environment from contaminants on handheld devices
- Easily accommodates aseptic technique
Recommended Storage
General Warehouse Storage - Keep out of direct sunlight
Specifications
Specifications
| For Use With (Equipment) | Smart Phone |
| Material | Polyethylene Film |
| Sterility | Sterile |
| Dimensions (L x W) | 22.9 x 35.6 cm (9 x 14 in.) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
