missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Wide Neck PFA Bottle
Bottles designed for long-term storage of solvents and standards (trace analytics).
Brand: BRAND™ 128764
Code : NEW
Additional Details : Weight : 0.34000kg
Description
- PFA screw cap with a molded sealing ring for a positive seal
- Autoclavable
- Easy to clean
- Withstands temperatures from -200 to 260°C
Specifications
2000 mL | |
PFA | |
Screw Cap |
Wide Neck | |
S40 |