missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Widefield Eyepieces with Crosshair Reticle

Product Code. 17138618
Click to view available options
Field of View:
20 mm
22 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17138618

Brand: Motic™ 1101001403641

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Widefield eyepiece for use with TEC POL Microscopes.

  • For use with Motic™ Panthera TEC POL Microscopes
  • 25 mm crosshair reticle
  • ±4 diopters

Specifications

Type Widefield Eyepiece
Color Black
Field of View 22 mm
For Use With (Equipment) Motic™ Panthera TEC POL Microscopes
Magnification Power 10X
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.