missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WIKI4
GREENER_CHOICE

Product Code. 18798446
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18798446

Brand: Tocris Bioscience™ 4855/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent tankyrase inhibitor

WIKI4 is an inhibitor of Wnt/β-catenin signaling (EC50 ∼ 75 nM). Inhibits auto-ADP-ribosylation of tankyrase 2 (TNKS2) (IC50 ∼15 nM); prevents the ubiquitination and degradation of axin. WIKI4 inhibits expression of β-catenin target genes in cancer cell lines as well as in human embryonic stem cells (ESC).

WIKI4 synthesized to Ancillary Material Grade also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione
CAS 838818-26-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C29H23N5O3S
Formula Weight Observed MW: 521.59
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.