missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ William's E Medium, GlutaMAX™ Supplement

GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

Brand:  Gibco™ 32551020

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10043282

  • 47.83 € / 500mL
Estimated Shipment: 08-08-2024
to see stock.

Description

Description

  • Developed by Williams and Gunn as reduced serum-supplemented medium for long-term cell cultures of adult rat liver epithelial cells
  • Gibco™ William's E Medium can also be used for growing hepatocyte cells of different species, such as human derived HepaRG™ cells
  • Medium contains unique ingredients including zinc, iron, manganese, non-essential amino acids, the reducing agent glutathione and the lipid methyl linoleate
  • GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format

Modified as follows

  • With: GlutaMAX™, phenol red
  • Without: HEPES

Using Willam's E Medium

  • Medium contains no proteins or growth factors therefore it requires supplementation, commonly with 5-10% Fetal Bovine Serum (FBS)
  • William's E Medium uses a sodium bicarbonate buffer system (2.2 g/L)
  • It requires a 5-10% CO2 environment to maintain physiological pH
Specifications

Specifications

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Mammalian Cell Culture
William's E Medium
No HEPES
GlutaMAX™
Room Temperature
Mammalian Cells
Liquid
GlutaMAX
Sustainable packaging
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers