Learn More
Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes
Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes fit tightly in any spinner turbine and have minimal NMR background.
Brand: Bel-Art™ 66000-0084
Code : NEW
Additional Details : Weight : 0.04310kg
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Description
Alert: Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.Specifications
Borosilicate Glass | |
4.947±0.019 mm | |
Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only. | |
8 in. | |
5/Pk. |
203 mm | |
100MHz | |
Tube and disposable cap | |
0.43 mm | |
Wilmad™ WG-5MM-ECONOMY-8-5 |