Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Finger for Varian/Bruker
Product Code. 15338628
Change view
Click to view available options
For Use With (Equipment):
Agilent/Varian NMR Spectrometer
Model:
UNI-FINGER-PEEK-3
Unit Size:
Each
Product Code. For Use With (Equipment) Model unitSize
15338628 Agilent/Varian NMR Spectrometer UNI-FINGER-PEEK-3 Each
1 options
Product Code. 15338628 Supplier Bel-Art™ SP Scienceware™ Supplier No. 660000055
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Finger for Varian/Bruker is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
  • Cost effective compared to the full VT version for sample changers
  • Allows a portion of the VT air to pass straight past the sample inside the spinner turbine therefore reducing VT gradients and micro sonic flutter due to a high VT flow
  • Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom.
  • Can also be used for spinning experiments.
Specifications
For Use With (Equipment) Agilent/Varian NMR Spectrometer
Model UNI-FINGER-PEEK-3