Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine
Product Code. 15368628
Change view
Click to view available options
For Use With (Equipment):
Agilent/Varian NMR Spectrometer
Model:
V-PEEK-3-NS
Unit Size:
Each
Product Code. For Use With (Equipment) Model unitSize
15368628 Agilent/Varian NMR Spectrometer V-PEEK-3-NS Each
1 options
Product Code. 15368628 Supplier Bel-Art™ SP Scienceware™ Supplier No. 660000058
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
  • Cost effective compared to the full VT version for sample changers
  • Allows a portion of the VT air to pass straight past the sample inside the spinner turbine therefore reducing VT gradients and micro sonic flutter due to a high VT flow
  • Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom.
  • Can also be used for spinning experiments.
Specifications
For Use With (Equipment) Agilent/Varian NMR Spectrometer
Includes PEEK exchangeable fingers
Model V-PEEK-3-NS