missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube is commonly used in sample degasification due to its capability to maintain vacuum quality.
42.00 € - 63.42 €
Product Code | Brand | Field Strength | Length (Metric) | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Field Strength | Length (Metric) | Model | Price | Quantity & Availability | |||
15388858
|
Bel-Art™ SP Scienceware™
66305-0571 |
300 MHz | 177.8 mm | 507-TR-7 |
42.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15308998
|
Bel-Art™ SP Scienceware™
66505-0575 |
500 MHz | 177.8 mm | 528-TR-7 |
46.90 €
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||
15318998
|
Bel-Art™ SP Scienceware™
66505-1095 |
400 MHz | 228.6 mm | 513-7TRA-9 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Each Screw-Cap Tube comes complete with open top screw cap and PTFE/Silicone septum
- Inert to most organic solvents and compounds but poor to strongly corrosive materials
- Remains reliable after multiple punctures
Specifications
5 mm | |
PTFE/Silicon Septum |