missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ WIN 18446

Product Code. 30001665
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001665 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001665 Supplier R&D Systems™ Supplier No. 4736/50
  • ONLINE EXCLUSIVE PRICE
    236.34 €
    212.10 € / 50mg
    Save 24.24 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. ALDH1a2 inhibitor

Inhibitor of aldehyde dehydrogenase 1a2 (ALDH1a2) (IC50 = 0.3 μM). Inhibits the biosynthesis of retinoic acid from retinol in neonatal and adult murine testis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1477-57-2
Quantity 50 mg
Product Line Tocris
Molecular Formula C12H20Cl4N2O2
Formula Weight 366.11
Purity >98%
Molecular Weight (g/mol) 366.104
Chemical Name or Material WIN 18446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.