Learn More
Contec Window Cleaning Squeegee and Replacement Parts
Product Code. 30280812 Shop All Contec Products
Change view
Click to view available options
Product Type:
Single-blade Squeegee
Unit Size:
Each
Product Code. Product Type Unit Size
30280812 Single-blade Squeegee Each
1 options
Product Code. 30280812 Supplier Contec Supplier No. WCSQ0018
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    150.21 € / Pack of 1
    Save 39.79 €
    20% Off
Estimated Shipment: 01-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel squeegee is lightweight, easy to use, and compatible with QuickConnect handles for hard-to-reach areas. Replacement rubber blade sold separately.
Features Benefits:
  • Lightweight, stainless-steel tool and rubber grip
  • Rubber grip prevents tool from slipping when used with disinfectants
  • Lightweight to clean tall windows and hard to reach areas
  • Maneuvers easily to use vertically and horizontally
Compatibility:
  • Compatible with all Contec's QuickConnect handles making windows easier to reach.
  • All components of the squeegee are compatible with common cleanroom cleaners and disinfectants
Press Down Handle Clip
  • Allows for easy installation preventing the rubber blade from moving during use
Screw-free channel
  • Blades to be replaced quickly and easily saving time
Autoclavable
  • Squeegee can be autoclaved at 121°C for 30 minutes
  • Ideal for use in all areas of the cleanroom
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single-blade Squeegee
Handle Material Stainless Steel
Autoclavable Autoclavable
Material Rubber