Learn More
Thermo Scientific™ Windows™ 7 OS Upgrade Kit for StepOne™ Real-Time PCR System
Product Code. 17216513
Change view
Click to view available options
Quantity:
2 Drives
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
17216513 2 Drives Pack of 2
1 options
Product Code. 17216513 Supplier Thermo Scientific™ Supplier No. A24637
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Allows you to reimage computers running Applied Biosystems™ real-time PCR data collection and analysis software to enable them to run using the Microsoft™ Windows™ 7 operating system (OS).
  • The Windows™ 7 OS Upgrade Kit for StepOne™ Real-Time PCR System allows you to reimage computers running Applied Biosystems™ real-time PCR data collection and analysis software to enable them to run using the Microsoft™ Windows™ 7 operating system (OS)
  • The kit includes the following: Recovery Image of Windows™ 7 64 Bit with SP1, 3 Partition, for Dell™ desktop OptiPlex™ XE only, Recovery Image of Windows™ 7 64 Bit with SP1, 3 Partition, for Dell™ Latitude™ E6520 only
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage OptiPlex XE Recovery Image, Windows 7 64 Bit SP1, 3 Partition, Latitude E6520 Recovery Image, Windows 7 64 Bit SP1, 3 Partition
For Use With (Equipment) StepOne™ System
Product Line Life Technologies
Software Type Computer Image
Quantity 2 Drives