Learn More
TERUMO Winged Infusion Sets SURFLO™
Product Code. 16417440
Change view
Click to view available options
Needle Gauge:
19 G
23 G
25 G
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
16417440 25 G Pack of 50
16497430 19 G Pack of 50
16407440 23 G Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16417440 Supplier TERUMO Supplier No. 8SV25NL30
  • ONLINE EXCLUSIVE PRICE
    71.72 €
    16.02 € / Pack of 50
    Save 55.70 €
    77% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Designed to ensure stability and grip during venipuncture
  • The Surflo™ Winged Infusion set is intended to introduce or withdraw fluids from the vascular system after vein puncture (18 - 27 G)
  • Highly transparent tubing and adapter for easy recognition of air bubbles and particles
  • The wings are large and flexible for a good grip and secure fixation
  • Reduced pressure on the blood vessel wall due to slightly angled wings
  • Suitable for all applications with a wide product range
TRUSTED_SUSTAINABILITY
Specifications
Product Type Infusion Set
Color Orange
Material PVC Tubing, Siliconized Needle
Needle Gauge 25 G
Sterility Sterile
Latex Free Yes