Learn More
Ansell™ Winter Monkey Grip™ 23-191 Series PVC Heavy Duty Gloves
Product Code. 12660782 Shop All Ansell Products
Change view
Click to view available options
Size:
10
11
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12660782 10 12 pairs
12670782 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12660782 Supplier Ansell™ Supplier No. 23191100
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Tough PVC vinyl coating protects hands from abrasions and stays soft and flexible in cold.
  • PVC coating repels water, oil and grease and remains flexible at low temperatures
  • Foam insulation keeps hands warm
  • Excellent grip for safety when handling heavy-duty objects
  • Curved fingers, wing thumb and soft jersey cotton lining offers superior comfort
  • Keeps hands warm, dry and protected from abrasions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Standards CATEGORY II EN 388 : 3121, EN 511 : 111, EN 1149, ANSI
Cuff Material PVC
Coating Material PVC
Color Beige
Cut Resistance Rating Level 1
Exterior Finish No Texture
Hand Specific
Material PVC
Powdered Powder Free
Product Type Heavy-duty Gloves
Size 10
Glove Cuff Style Knit
Liner Material Cotton Jersey
Lining Knit Lining
Palm Type Fully coated-palm
Quantity 72 Pairs
Resistant To Oil and grease repellency
Series 23-191