Learn More
Ansell™ Winter Monkey Grip™ 23-191 Series PVC Heavy Duty Gloves
Product Code. p-8005402
Click to view available options
Size:
10
11
Unit Size:
12 pairs
Product Code. 12660782
Brand: Ansell™ 23191100
Only null left
Add to basket
Edge
Add to basket
Tough PVC vinyl coating protects hands from abrasions and stays soft and flexible in cold.
  • PVC coating repels water, oil and grease and remains flexible at low temperatures
  • Foam insulation keeps hands warm
  • Excellent grip for safety when handling heavy-duty objects
  • Curved fingers, wing thumb and soft jersey cotton lining offers superior comfort
  • Keeps hands warm, dry and protected from abrasions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Standards CATEGORY II EN 388 : 3121, EN 511 : 111, EN 1149, ANSI
Cuff Material PVC
Coating Material PVC
Color Beige
Cut Resistance Rating Level 1
Exterior Finish No Texture
Hand Specific
Material PVC
Powdered Powder Free
Product Type Heavy-duty Gloves
Size 10
Glove Cuff Style Knit
Liner Material Cotton Jersey
Lining Knit Lining
Palm Type Fully coated-palm
Quantity 72 Pairs
Resistant To Oil and grease repellency
Series 23-191