missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios WIP'Anios Excel

Product Code. 15398954
Change view
Click to view available options
Packaging Quantity:
12 Pk.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
15398954 12 Pk. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15398954 Supplier Laboratoires Anios Supplier No. 2446424DI
  • ONLINE EXCLUSIVE PRICE
    145.50 €
    122.82 € / Pack of 12
    Save 22.68 €
    15% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Surface treatment, non-submersible and non-invasive medical devices, and wiping of endoscopy sheaths in preliminary treatment, prior to submersion for cleaning and disinfection.

  • 100% Biodegradable viscose wipes (only Wip’Anios Excel)
    Impregnating solution:
  • Formulation fragrance and alcohol free
  • Broad virucidal spectrum
  • Anti-microbial efficacy from 2 minutes

Specifications

Product Type WIP'Anios Excel
For Use With (Application) For Cleaning and Pre Disinfection of Medical Surgical Instrumentation, Medical Devices, Heat Sensitive Instruments and Endoscopy Equipment
Packaging Quantity 12 Pk.
compliance-icons
Product Identifier
  • WIP'ANIOS EXCEL
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: DIDECYLDIMETHYLAMMONIUM CHLORIDE, N-C12-14-ALKYLTRIMETHYLENEDI-AMINES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.