missing translation for 'onlineSavingsMsg'
Learn More

AZOWIPE™ Wipes

Product Code. 12388667
Change view
Click to view available options
Dimensions (L x W):
220 x 200 mm
Unit Size:
0.25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12388667 220 x 200 mm 0.25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12388667 Supplier Vernacare Supplier No. 81103

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bactericidal hard surface disinfectant wipes. AZOWIPE™ Wipes are convenient for use in all clinical environments and are an important part of a robust infection control protocol.

  • Made from a viscose rayon nonwoven material strong and durable in use
  • Foil sealed - prevents evaporation in storage
  • Suitable for all hard surfaces - one product from a single source
  • Contains NO preservative - no smearing or streaking when in use
  • Specially designed canister - easy to dispense wipes
  • 70% Solution of Isopropyl Alcohol BP
  • AZOWIPE - for the disinfection of hard surfaces
  • Dimensions: 200mm x 220mm

Specifications

Product Type Wipes
Material Viscose Rayon
Dimensions (L x W) 220 x 200 mm
compliance-icons compliance-icons
Product Identifier
  • AZOWIPE Equipment Wipes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.